Support
Deals
chevron-down
Special Offers
Spotlight
Product of the Month
Outlet Center
View All
Sign In
chevron-down
chevron-down
Fisher Scientific
Centrifuge guide
Centrifuge applications tools
RPM / RCF Calculator
RPM / RCF Calculator
Relative Centrifugal Force (RCF)
Revolutions per minute of rotor (rpm)
Radius of rotor (mm)
Relative Centrifugal Force (RCF)
Revolutions per minute (rpm)
Relative Centrifugal Force (RCF)
Radius of rotor (mm)
Revolutions per minute of rotor (rpm)
RCF = 1.12 x Radius x (rpm/1000)
2